Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Methylbenzotriazole

Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA3059909

 
alfa aesar™
30599-09
10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA3059918

 
alfa aesar™
30599-18
50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Quantity 10g
Melting Point 72° to 75°C
Quantity 10g
Chemical Name or Material 5-Methylbenzotriazole
CAS 136-85-6
Physical Form Crystalline
Formula Weight 133.14g/mol
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H311-H302-H315-H319-H335
Toxic in contact with skin.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P361-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several min
TSCA TSCA
Safety and Handling

Danger

GHS P Statement
P261-P280-P305+P351+P338-P361-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several min

GHS H Statement
H311-H302-H315-H319-H335
Toxic in contact with skin.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.