missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Alfa Aesar™ 5-Methylbenzofuroxan, 97%
Manufacturer: Alfa Aesar™ A1512406
Specifications
5-Methylbenzofuroxan | |
96°C to 98°C | |
19164-41-1 | |
MFCD00068057 | |
5-methylbenzofuroxan, benzofurazan, 5-methyl-, 1-oxide, 5-methylbenzofurazan-1-oxide, 5-methyl-2,1,3-benzoxadiazol-1-ium-1-olate, 5-methylbenzofurazan oxide, 2,1,3-benzoxadiazole, 5-methyl-, monooxide, 5-methylbenzofurazan 1-oxide, acmc-20am21, 5-methylbenzfurazan-1-oxide, 5-methylbenzofurazan n-oxide | |
CC1=CC2=NO[N+](=C2C=C1)[O-] | |
150.137 | |
150.15 |
Odorless | |
5g | |
C7H6N2O2 | |
137386 | |
HCWVKDOCXDWFEH-UHFFFAOYSA-N | |
5-methyl-1-oxido-2,1,3-benzoxadiazol-1-ium | |
140486 | |
97% |
Chemical Identifiers
19164-41-1 | |
150.137 | |
HCWVKDOCXDWFEH-UHFFFAOYSA-N | |
140486 | |
CC1=CC2=NO[N+](=C2C=C1)[O-] |
C7H6N2O2 | |
MFCD00068057 | |
5-methylbenzofuroxan, benzofurazan, 5-methyl-, 1-oxide, 5-methylbenzofurazan-1-oxide, 5-methyl-2,1,3-benzoxadiazol-1-ium-1-olate, 5-methylbenzofurazan oxide, 2,1,3-benzoxadiazole, 5-methyl-, monooxide, 5-methylbenzofurazan 1-oxide, acmc-20am21, 5-methylbenzfurazan-1-oxide, 5-methylbenzofurazan n-oxide | |
5-methyl-1-oxido-2,1,3-benzoxadiazol-1-ium |