missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Methylbenzofuroxan, 97%

Manufacturer:  Alfa Aesar™ A1512403

 View more versions of this product

Catalog No. AAA1512403



Specifications

Specifications

5-Methylbenzofuroxan
96°C to 98°C
19164-41-1
MFCD00068057
5-methylbenzofuroxan, benzofurazan, 5-methyl-, 1-oxide, 5-methylbenzofurazan-1-oxide, 5-methyl-2,1,3-benzoxadiazol-1-ium-1-olate, 5-methylbenzofurazan oxide, 2,1,3-benzoxadiazole, 5-methyl-, monooxide, 5-methylbenzofurazan 1-oxide, acmc-20am21, 5-methylbenzfurazan-1-oxide, 5-methylbenzofurazan n-oxide
CC1=CC2=NO[N+](=C2C=C1)[O-]
150.137
150.15
Odorless
1g
C7H6N2O2
137386
HCWVKDOCXDWFEH-UHFFFAOYSA-N
5-methyl-1-oxido-2,1,3-benzoxadiazol-1-ium
140486
97%

Chemical Identifiers

19164-41-1
150.137
HCWVKDOCXDWFEH-UHFFFAOYSA-N
140486
CC1=CC2=NO[N+](=C2C=C1)[O-]
C7H6N2O2
MFCD00068057
5-methylbenzofuroxan, benzofurazan, 5-methyl-, 1-oxide, 5-methylbenzofurazan-1-oxide, 5-methyl-2,1,3-benzoxadiazol-1-ium-1-olate, 5-methylbenzofurazan oxide, 2,1,3-benzoxadiazole, 5-methyl-, monooxide, 5-methylbenzofurazan 1-oxide, acmc-20am21, 5-methylbenzfurazan-1-oxide, 5-methylbenzofurazan n-oxide
5-methyl-1-oxido-2,1,3-benzoxadiazol-1-ium
Documents