missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

5-Methyl N-Carbobenzoxy-L-glutamate, TCI America™

$87.08 - $291.81

Chemical Identifiers

CAS 4652-65-7
Molecular Formula C14H17NO6
Molecular Weight (g/mol) 295.291
MDL Number MFCD00191090
InChI Key JSNVOAWDAFVIKY-NSHDSACASA-N
Synonym 5-Methyl N-Cbz-L-glutamate, N-Carbobenzoxy-L-glutamic Acid 5-Methyl Ester, N-Cbz-L-glutamic Acid 5-Methyl Ester, Z-Glu(OMe)-OH
PubChem CID 6350584
IUPAC Name (2S)-5-methoxy-5-oxo-2-(phenylmethoxycarbonylamino)pentanoic acid
SMILES COC(=O)CCC(C(=O)O)NC(=O)OCC1=CC=CC=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M226925G View Documents TCI America
M226925G
25g Each for $291.81
 
M22695G View Documents TCI America
M22695G
5g Each for $87.08
 
Specifications

Chemical Identifiers

4652-65-7
295.291
JSNVOAWDAFVIKY-NSHDSACASA-N
6350584
COC(=O)CCC(C(=O)O)NC(=O)OCC1=CC=CC=C1
C14H17NO6
MFCD00191090
5-Methyl N-Cbz-L-glutamate, N-Carbobenzoxy-L-glutamic Acid 5-Methyl Ester, N-Cbz-L-glutamic Acid 5-Methyl Ester, Z-Glu(OMe)-OH
(2S)-5-methoxy-5-oxo-2-(phenylmethoxycarbonylamino)pentanoic acid

Specifications

White
25g
C14H17NO6
5-Methyl N-Cbz-L-glutamate, N-Carbobenzoxy-L-glutamic Acid 5-Methyl Ester, N-Cbz-L-glutamic Acid 5-Methyl Ester, Z-Glu(OMe)-OH
COC(=O)CCC(C(=O)O)NC(=O)OCC1=CC=CC=C1
295.291
295.29
≥98.0% (HPLC,T)
70°C
4652-65-7
MFCD00191090
JSNVOAWDAFVIKY-NSHDSACASA-N
(2S)-5-methoxy-5-oxo-2-(phenylmethoxycarbonylamino)pentanoic acid
6350584
Crystal-Powder at 20°C
5-Methyl N-Carbobenzoxy-L-glutamate
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Refrigerator