Please login to your online account to display your discounted pricing

5-Methyl-4-isoxazolecarbonyl chloride, 97%, Maybridge


Quantity 5g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  CC26302DE

 View more versions of this product

Catalog No. CC26302DE

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 145.54
Quantity 5g
Assay 97%
Chemical Name or Material 5-Methyl-4-isoxazolecarbonyl chloride
Packaging Amber Glass Bottle
Assay Percent Range 97%
Molecular Formula C5H4ClNO2
CAS 67305-24-2

Specifications

Chemical Name or Material 5-Methyl-4-isoxazolecarbonyl chloride
Assay 97%
Assay Percent Range 97%
Molecular Formula C5H4ClNO2
Formula Weight 145.54