missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Methyl-3-trifluoromethyl-1H-pyrazole, 98%

$30.99 - $300.72

Chemical Identifiers

CAS 10010-93-2
Molecular Formula C5H5F3N2
Molecular Weight (g/mol) 150.104
MDL Number MFCD00156058
InChI Key DLCHCAYDSKIFIN-UHFFFAOYSA-N
Synonym 5-methyl-3-trifluoromethyl-1h-pyrazole, 3-methyl-5-trifluoromethyl-1h-pyrazole, 3-methyl-5-trifluoromethyl pyrazole, 1h-pyrazole, 3-methyl-5-trifluoromethyl, 5-methyl-3-trifluoromethyl pyrazole, 3-trifluoromethyl-5-methyl pyrazole, pyrazole, 3-methyl-5-trifluoromethyl, 3-methyl-5-trifluoromethylpyrazole, 3-trifluoromethyl-5-methylpyrazole, 3-methyl-5-trifluoromethyl-2h-pyrazole
PubChem CID 139077
IUPAC Name 5-methyl-3-(trifluoromethyl)-1H-pyrazole
SMILES CC1=CC(=NN1)C(F)(F)F
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH64308MD View Documents Alfa Aesar™
H64308MD
250mg Each for $30.99
 
AAH6430803 View Documents Alfa Aesar™
H6430803
1g Each for $64.00
 
AAH6430806 View Documents Alfa Aesar™
H6430806
5g Each for $300.72
 
Specifications

Chemical Identifiers

10010-93-2
150.104
DLCHCAYDSKIFIN-UHFFFAOYSA-N
139077
CC1=CC(=NN1)C(F)(F)F
C5H5F3N2
MFCD00156058
5-methyl-3-trifluoromethyl-1h-pyrazole, 3-methyl-5-trifluoromethyl-1h-pyrazole, 3-methyl-5-trifluoromethyl pyrazole, 1h-pyrazole, 3-methyl-5-trifluoromethyl, 5-methyl-3-trifluoromethyl pyrazole, 3-trifluoromethyl-5-methyl pyrazole, pyrazole, 3-methyl-5-trifluoromethyl, 3-methyl-5-trifluoromethylpyrazole, 3-trifluoromethyl-5-methylpyrazole, 3-methyl-5-trifluoromethyl-2h-pyrazole
5-methyl-3-(trifluoromethyl)-1H-pyrazole

Specifications

White
250mg
C5H5F3N2
743044
DLCHCAYDSKIFIN-UHFFFAOYSA-N
5-methyl-3-(trifluoromethyl)-1H-pyrazole
139077
Powder
5-Methyl-3-trifluoromethyl-1H-pyrazole
88°C to 90°C
10010-93-2
MFCD00156058
5-methyl-3-trifluoromethyl-1h-pyrazole, 3-methyl-5-trifluoromethyl-1h-pyrazole, 3-methyl-5-trifluoromethyl pyrazole, 1h-pyrazole, 3-methyl-5-trifluoromethyl, 5-methyl-3-trifluoromethyl pyrazole, 3-trifluoromethyl-5-methyl pyrazole, pyrazole, 3-methyl-5-trifluoromethyl, 3-methyl-5-trifluoromethylpyrazole, 3-trifluoromethyl-5-methylpyrazole, 3-methyl-5-trifluoromethyl-2h-pyrazole
CC1=CC(=NN1)C(F)(F)F
150.104
150.1
98%
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning