Please login to your online account to display your discounted pricing

5-Methyl-3-phenyl-4-isoxazolamine, 97%, Maybridge

Chemical Identifiers

Chemical Name or Material 5-Methyl-3-phenyl-4-isoxazolamine
CAS 21169-65-3
CAS Min % 97.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C10H10N2O
Formula Weight 174.2
MDL Number MFCD02677726
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

TL01028CB

 
fisher scientific company
TL01028CB
Amber Glass Bottle 250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 174.2
Quantity 250mg
Assay 97%
CAS Min % 97.0
Chemical Name or Material 5-Methyl-3-phenyl-4-isoxazolamine
Molecular Formula C10H10N2O
CAS 21169-65-3
Health Hazard 2 GHS P Statement Causes skin irritation. May cause respiratory irritation. Causes serious eye irritation.
Packaging Amber Glass Bottle
Assay Percent Range 97%
MDL Number MFCD02677726
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement IF ON SKIN: Wash with plenty of soap and water. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. IF INHALED: Remove to fr