Please login to your online account to display your discounted pricing

5-Methyl-3-phenyl-4-isoxazolamine, 97%, Maybridge


Quantity 250mg
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  TL01028CB

 View more versions of this product

Catalog No. TL01028CB

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 174.2
Quantity 250mg
Assay 97%
CAS Min % 97.0
Chemical Name or Material 5-Methyl-3-phenyl-4-isoxazolamine
Molecular Formula C10H10N2O
CAS 21169-65-3
Health Hazard 2 GHS P Statement Causes skin irritation. May cause respiratory irritation. Causes serious eye irritation.
Packaging Amber Glass Bottle
Assay Percent Range 97%
MDL Number MFCD02677726
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement IF ON SKIN: Wash with plenty of soap and water. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. IF INHALED: Remove to fr

Specifications

Chemical Name or Material 5-Methyl-3-phenyl-4-isoxazolamine
CAS 21169-65-3
CAS Min % 97.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C10H10N2O
Formula Weight 174.2
MDL Number MFCD02677726
Accessories

Warning

GHS P Statement IF ON SKIN: Wash with plenty of soap and water. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. IF INHALED: Remove to fr

GHS P Statement Causes skin irritation. May cause respiratory irritation. Causes serious eye irritation.

SDS & Safety Documents