missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Methyl-3-isoxazolemethanol, 97%

Manufacturer:  Alfa Aesar™ H6194803

 View more versions of this product

Catalog No. AAH6194803


 

Specifications

Specifications

5-Methyl-3-isoxazolemethanol
97%
MFCD00085129
1g
CC1=CC(=NO1)CO
113.116
113.11
35166-33-7
C5H7NO2
Colorless
MDYHWQQHEWDJKR-UHFFFAOYSA-N
(5-methyl-1,2-oxazol-3-yl)methanol
2736567
Liquid

Chemical Identifiers

35166-33-7
113.116
MDYHWQQHEWDJKR-UHFFFAOYSA-N
C5H7NO2
MFCD00085129
2736567
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

SDS Documents