Please login to your online account to display your discounted pricing

5-methyl-3-isoxazolecarbaldehyde, 97%, Maybridge

$121.74 - $121.74

Chemical Identifiers

Chemical Name or Material 5-Methyl-3-isoxazolecarbaldehyde
Assay 97%
Assay Percent Range 97%
Molecular Formula C5H5NO2
Formula Weight 111.1
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

CC10404DE

  SDS
fisher scientific company
CC10404DE
Amber Glass Bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CC10404DA

  SDS
fisher scientific company
CC10404DA
Amber Glass Bottle 1g Each for $121.74
Description & Specifications

Specifications

Formula Weight 111.1
Quantity 5g
Assay 97%
Chemical Name or Material 5-Methyl-3-isoxazolecarbaldehyde
Packaging Amber Glass Bottle
Assay Percent Range 97%
Molecular Formula C5H5NO2
CAS 62254-74-4