Please login to your online account to display your discounted pricing

5-methyl-3-isoxazolecarbaldehyde, 97%, Maybridge


Quantity 1g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  CC10404DA

 View more versions of this product

Catalog No. CC10404DA

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 111.1
Quantity 1g
Assay 97%
Chemical Name or Material 5-Methyl-3-isoxazolecarbaldehyde
CAS 62254-74-4
Packaging Amber Glass Bottle
Assay Percent Range 97%
Molecular Formula C5H5NO2

Specifications

Chemical Name or Material 5-Methyl-3-isoxazolecarbaldehyde
Assay 97%
Assay Percent Range 97%
Molecular Formula C5H5NO2
Formula Weight 111.1