missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

5-Methyl-3-heptene (cis- and trans- mixture), TCI America™

Manufacturer:  TCI America M09931ML

 View more versions of this product

Catalog No. M09931ML



Specifications

Specifications

5-Methyl-3-heptene (cis- and trans- mixture)
1mL
C8H16
3295
CCC=CC(C)CC
112.216
112.22
≥98.0% (GC)
Yellow
13172-91-3
MFCD00049124
YMNTZRCUPAYGLG-UHFFFAOYSA-N
5-methylhept-3-ene
143054
Clear Liquid at 20°C

Chemical Identifiers

13172-91-3
112.216
YMNTZRCUPAYGLG-UHFFFAOYSA-N
5-methylhept-3-ene
C8H16
MFCD00049124
143054
CCC=CC(C)CC
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Refrigerator

Documents