missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

5-Methyl-3-heptene (cis- and trans- mixture), TCI America™

$406.39 - $696.69

Chemical Identifiers

CAS 13172-91-3
Molecular Formula C8H16
Molecular Weight (g/mol) 112.216
MDL Number MFCD00049124
InChI Key YMNTZRCUPAYGLG-UHFFFAOYSA-N
PubChem CID 143054
IUPAC Name 5-methylhept-3-ene
SMILES CCC=CC(C)CC
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M09931ML View Documents TCI America
M09931ML
1mL Each for $406.39
 
M09935ML View Documents TCI America
M09935ML
5mL Each for $696.69
 
Specifications

Chemical Identifiers

13172-91-3
112.216
YMNTZRCUPAYGLG-UHFFFAOYSA-N
5-methylhept-3-ene
C8H16
MFCD00049124
143054
CCC=CC(C)CC

Specifications

Yellow
13172-91-3
MFCD00049124
YMNTZRCUPAYGLG-UHFFFAOYSA-N
5-methylhept-3-ene
143054
Clear Liquid at 20°C
5-Methyl-3-heptene (cis- and trans- mixture)
1mL
C8H16
3295
CCC=CC(C)CC
112.216
112.22
≥98.0% (GC)
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Refrigerator