missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Methyl-3-(4-pyridyl)-1,2,4-oxadiazole, 97%

Manufacturer:  Alfa Aesar™ H54416MD

 View more versions of this product

Catalog No. AAH54416MD



Specifications

Specifications

5-Methyl-3-(4-pyridyl)-1,2,4-oxadiazole
10350-70-6
MFCD00464110
JZIKSOZOBQKFEX-UHFFFAOYSA-N
5-methyl-3-pyridin-4-yl-1,2,4-oxadiazole
2771672
97%
250mg
C8H7N3O
4-5-methyl-1,2,4-oxadiazol-3-yl pyridine
CC1=NC(=NO1)C2=CC=NC=C2
161.164
161.16

Chemical Identifiers

10350-70-6
161.164
JZIKSOZOBQKFEX-UHFFFAOYSA-N
2771672
CC1=NC(=NO1)C2=CC=NC=C2
C8H7N3O
MFCD00464110
4-5-methyl-1,2,4-oxadiazol-3-yl pyridine
5-methyl-3-pyridin-4-yl-1,2,4-oxadiazole
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

Documents