missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Methyl-3-(4-methylphenyl)-1,2,4-oxadiazole, 97%

Manufacturer:  Alfa Aesar™ H5439906

 View more versions of this product

Catalog No. AAH5439906



Specifications

Specifications

5-Methyl-3-(4-methylphenyl)-1,2,4-oxadiazole
81386-30-3
MFCD00085801
3-4-methylphenyl-5-methyl-1,2,4-oxadiazole
CC1=CC=C(C=C1)C2=NOC(=N2)C
174.203
174.2
5g
C10H10N2O
7438
OYGSEQYNSZOSCE-UHFFFAOYSA-N
5-methyl-3-(4-methylphenyl)-1,2,4-oxadiazole
270750
97%

Chemical Identifiers

81386-30-3
174.203
OYGSEQYNSZOSCE-UHFFFAOYSA-N
270750
CC1=CC=C(C=C1)C2=NOC(=N2)C
C10H10N2O
MFCD00085801
3-4-methylphenyl-5-methyl-1,2,4-oxadiazole
5-methyl-3-(4-methylphenyl)-1,2,4-oxadiazole
Safety and Handling

Safety and Handling

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

Documents