missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

5-Methyl-3,4-diphenylisoxazole, TCI America™

$91.67 - $316.25

Chemical Identifiers

CAS 37928-17-9
Molecular Formula C16H13NO
Molecular Weight (g/mol) 235.286
MDL Number MFCD18449672
InChI Key ZXIRUKJWLADSJS-UHFFFAOYSA-N
Synonym 5-methyl-3,4-diphenylisoxazole, 5-methyl-3,4-diphenyl-isoxazole, isoxazole, 5-methyl-3,4-diphenyl, d04ypp, 3,4-diphenyl-5-methylisoxazole, 3,4-diphenyl-5-methyl isoxazole, 3,4-diphenyl-5-methyl-isoxazole, 5-methyl-3,4-diphenyl isoxazole
PubChem CID 9991673
IUPAC Name 5-methyl-3,4-diphenyl-1,2-oxazole
SMILES CC1=C(C(=NO1)C2=CC=CC=C2)C3=CC=CC=C3
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M28301G View Documents TCI America
M28301G
1g Each for $91.67
 
M28305G View Documents TCI America
M28305G
5g Each for $316.25
 
Specifications

Chemical Identifiers

37928-17-9
235.286
ZXIRUKJWLADSJS-UHFFFAOYSA-N
9991673
CC1=C(C(=NO1)C2=CC=CC=C2)C3=CC=CC=C3
C16H13NO
MFCD18449672
5-methyl-3,4-diphenylisoxazole, 5-methyl-3,4-diphenyl-isoxazole, isoxazole, 5-methyl-3,4-diphenyl, d04ypp, 3,4-diphenyl-5-methylisoxazole, 3,4-diphenyl-5-methyl isoxazole, 3,4-diphenyl-5-methyl-isoxazole, 5-methyl-3,4-diphenyl isoxazole
5-methyl-3,4-diphenyl-1,2-oxazole

Specifications

White-Yellow
1g
C16H13NO
5-methyl-3,4-diphenylisoxazole, 5-methyl-3,4-diphenyl-isoxazole, isoxazole, 5-methyl-3,4-diphenyl, d04ypp, 3,4-diphenyl-5-methylisoxazole, 3,4-diphenyl-5-methyl isoxazole, 3,4-diphenyl-5-methyl-isoxazole, 5-methyl-3,4-diphenyl isoxazole
CC1=C(C(=NO1)C2=CC=CC=C2)C3=CC=CC=C3
235.286
235.29
≥98.0% (GC)
98°C
37928-17-9
MFCD18449672
ZXIRUKJWLADSJS-UHFFFAOYSA-N
5-methyl-3,4-diphenyl-1,2-oxazole
9991673
Crystal-Powder at 20°C
5-Methyl-3,4-diphenylisoxazole
Safety and Handling

Safety and Handling

TSCA : No