Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Methyl-2-(trifluoromethyl)benzoic acid, 97%

$190.86 - $574.15

Chemical Identifiers

Chemical Name or Material 5-Methyl-2-(trifluoromethyl)benzoic acid
CAS 120985-68-4
Molecular Formula C9H7F3O2
Formula Weight 204.15
MDL Number MFCD06660291
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2637706

  SDS
alfa aesar™
H2637706
5g Each for $574.15

AAH2637703

  SDS
alfa aesar™
H2637703
1g Each for $190.86
Description & Specifications

Specifications

Formula Weight 204.15
Chemical Name or Material 5-Methyl-2-(trifluoromethyl)benzoic acid
Assay 97%
Molecular Formula C9H7F3O2
MDL Number MFCD06660291
CAS 120985-68-4
Health Hazard 1 Warning
Melting Point 90° to 94°C
Quantity 5g
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Solubility
Soluble in alcohol.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.