Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Methyl-2-(trifluoromethyl)benzaldehyde, 97%

$272.45 - $821.97

Chemical Identifiers

Chemical Name or Material 5-Methyl-2-(trifluoromethyl)benzaldehyde
CAS 886502-76-7
Molecular Formula C9H7F3O
Formula Weight 188.15
MDL Number MFCD06660294
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2641106

 
alfa aesar™
H2641106
5g Each for $821.97

AAH2641103

 
alfa aesar™
H2641103
1g Each for $272.45
Description & Specifications

Specifications

Formula Weight 188.15
Chemical Name or Material 5-Methyl-2-(trifluoromethyl)benzaldehyde
Assay 97%
Molecular Formula C9H7F3O
MDL Number MFCD06660294
CAS 886502-76-7
Health Hazard 1 Warning
Quantity 5g
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Applications
5-Methyl-2-(trifluoromethyl)benzaldehyde

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents. It is sensitive to air. Store under dry inert gas.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.