Please login to your online account to display your discounted pricing

[5-Methyl-2-(trifluoromethyl)-3-furyl]methylamine, 97%, Maybridge


Quantity 5g

Manufacturer: fisher scientific company  CC00713DE

 View more versions of this product

Catalog No. CC00713DE

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 179.14
Quantity 5g
Assay 97%
Chemical Name or Material [5-Methyl-2-(trifluoromethyl)-3-furyl]methylamine
Assay Percent Range 97%
Molecular Formula C7H8F3NO
CAS 306935-05-7

Specifications

Chemical Name or Material [5-Methyl-2-(trifluoromethyl)-3-furyl]methylamine
Assay 97%
Assay Percent Range 97%
Molecular Formula C7H8F3NO
Formula Weight 179.14