missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
[5-Methyl-2-(trifluoromethyl)-3-furyl]methylamine, 97%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals CC00713DA
Specifications
[5-Methyl-2-(trifluoromethyl)-3-furyl]methylamine | |
97% | |
C7H8F3NO | |
1 g | |
ROYYYTOVBUUPDX-UHFFFAOYSA-N | |
[5-methyl-2-(trifluoromethyl)furan-3-yl]methanamine | |
2779900 | |
97% |
306935-05-7 | |
Amber Glass Bottle | |
MFCD02180792 | |
5-methyl-2-trifluoromethyl-3-furyl methylamine, 5-methyl-2-trifluoromethyl furan-3-yl methanamine, 3-aminomethyl-5-methyl-2-trifluoromethyl furan, 1-5-methyl-2-trifluoromethyl furan-3-yl methanamine, 5-methyl-2-trifluoromethyl-3-furanyl methanamine | |
CC1=CC(CN)=C(O1)C(F)(F)F | |
179.14 | |
179.14 |
Chemical Identifiers
306935-05-7 | |
179.14 | |
ROYYYTOVBUUPDX-UHFFFAOYSA-N | |
2779900 | |
CC1=CC(CN)=C(O1)C(F)(F)F |
C7H8F3NO | |
MFCD02180792 | |
5-methyl-2-trifluoromethyl-3-furyl methylamine, 5-methyl-2-trifluoromethyl furan-3-yl methanamine, 3-aminomethyl-5-methyl-2-trifluoromethyl furan, 1-5-methyl-2-trifluoromethyl furan-3-yl methanamine, 5-methyl-2-trifluoromethyl-3-furanyl methanamine | |
[5-methyl-2-(trifluoromethyl)furan-3-yl]methanamine |