Please login to your online account to display your discounted pricing

5-Methyl-2-(trifluoromethyl)-3-furyl isocyanate, 97%, Maybridge

Chemical Identifiers

Chemical Name or Material 5-Methyl-2-(trifluoromethyl)-3-furyl isocyanate
Assay 90%
Assay Percent Range 90%
Molecular Formula C7H4F3NO2
Formula Weight 191.11
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

CC00706DE

  SDS
fisher scientific company
CC00706DE
Amber Glass Bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CC00706CB

  SDS
fisher scientific company
CC00706CB
Amber Glass Bottle 250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CC00706DA

  SDS
fisher scientific company
CC00706DA
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 191.11
Quantity 5g
Assay 90%
Packaging Amber Glass Bottle
Molecular Formula C7H4F3NO2
Chemical Name or Material 5-Methyl-2-(trifluoromethyl)-3-furyl isocyanate
Assay Percent Range 90%
CAS 306935-03-5