Please login to your online account to display your discounted pricing

5-Methyl-2-(trifluoromethyl)-3-furoic acid, 97%, Maybridge


Quantity 250mg
Packaging Amber glass bottle

Manufacturer: fisher scientific company  CC00701CB

 View more versions of this product

Catalog No. CC00701CB

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 194.11
Quantity 250mg
Assay 97%
Assay Percent Range 97%
Chemical Name or Material 5-Methyl-2-(trifluoromethyl)-3-furoic acid
Packaging Amber glass bottle
Molecular Formula C7H5F3O3
CAS 17515-74-1

Specifications

Chemical Name or Material 5-Methyl-2-(trifluoromethyl)-3-furoic acid
Assay 97%
Assay Percent Range 97%
Molecular Formula C7H5F3O3
Formula Weight 194.11