missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

5-Methyl-2-thiouracil, TCI America™

Manufacturer:  TCI America M099410G

 View more versions of this product

Catalog No. M099410G



Specifications

Specifications

5-Methyl-2-thiouracil
10g
C5H6N2OS
4-Hydroxy-2-mercapto-5-methylpyrimidine, 4-Hydroxy-5-methyl-2-thiopyrimidine
CC1=CNC(=S)NC1=O
142.176
142.18
≥98.0% (T)
White
636-26-0
MFCD00009777
ZLAQATDNGLKIEV-UHFFFAOYSA-N
5-methyl-2-sulfanylidene-1H-pyrimidin-4-one
720471
Crystal-Powder at 20°C

Chemical Identifiers

636-26-0
142.176
ZLAQATDNGLKIEV-UHFFFAOYSA-N
720471
CC1=CNC(=S)NC1=O
C5H6N2OS
MFCD00009777
4-Hydroxy-2-mercapto-5-methylpyrimidine, 4-Hydroxy-5-methyl-2-thiopyrimidine
5-methyl-2-sulfanylidene-1H-pyrimidin-4-one
Safety and Handling

Safety and Handling

RTECSNumber : XP2108500

TSCA : Yes

Documents