Please login to your online account to display your discounted pricing

5-Methyl-2-thiophenecarboxylic acid, 97%

Chemical Identifiers

Chemical Name or Material 5-Methyl-2-thiophenecarboxylic acid
CAS 1918-79-2
CAS Min % 98.5
CAS Max % 100.0
Assay 98.5 to 101.5% (Acid)
Assay Percent Range 97%
Molecular Formula C6H6O2S
Formula Weight 142.18
MDL Number MFCD00005439
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC12788EA

  SDS
fisher scientific company
AC12788EA
Amber Glass Bottle 10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Liquid
Assay 98.5 to 101.5% (Acid)
Formula Weight 142.18
Melting Point 136° to 139°C
Quantity 10g
Color Clear
CAS Max % 100.0
MDL Number MFCD00005439
CAS Min % 98.5
Chemical Name or Material 5-Methyl-2-thiophenecarboxylic acid
Infrared Spectrum Authentic
Packaging Amber Glass Bottle
Assay Percent Range 97%
Molecular Formula C6H6O2S
CAS 1918-79-2
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.