missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
5-methyl-2-phenyl-2H-1,2,3-triazole-4-carbonyl chloride, Thermo Scientific™
Supplier: Thermo Scientific Chemicals CC16202CB
Specifications
5-Methyl-2-phenyl-2H-1, 2, 3-triazole-4-carbonyl chloride | |
97% | |
C10H8ClN3O | |
250 mg | |
UJYBUZMRRLFXGM-UHFFFAOYSA-N | |
5-methyl-2-phenyltriazole-4-carbonyl chloride | |
2776447 | |
97% |
36401-55-5 | |
Amber Glass Bottle | |
MFCD00052545 | |
5-methyl-2-phenyl-2h-1,2,3-triazole-4-carbonyl chloride, 5-methyl-2-phenyl-1,2,3-triazole-4-carbonyl chloride, 2h-1,2,3-triazole-4-carbonylchloride, 5-methyl-2-phenyl, 5-methyl-2-phenyl-2h-1,2,3 triazole-4-carbonyl chloride, 5-methyl-2-phenyl-4-triazolecarbonyl chloride, 5-methyl-2-phenyl-2h-1,2,3 triazole-4-carbo nyl chloride, 5-methyl-2-phenyl-2h-1,2,3-triazole-4-carbonylchloride | |
CC1=NN(N=C1C(Cl)=O)C1=CC=CC=C1 | |
221.64 | |
221.65 |
Chemical Identifiers
36401-55-5 | |
221.64 | |
UJYBUZMRRLFXGM-UHFFFAOYSA-N | |
2776447 | |
CC1=NN(N=C1C(Cl)=O)C1=CC=CC=C1 |
C10H8ClN3O | |
MFCD00052545 | |
5-methyl-2-phenyl-2h-1,2,3-triazole-4-carbonyl chloride, 5-methyl-2-phenyl-1,2,3-triazole-4-carbonyl chloride, 2h-1,2,3-triazole-4-carbonylchloride, 5-methyl-2-phenyl, 5-methyl-2-phenyl-2h-1,2,3 triazole-4-carbonyl chloride, 5-methyl-2-phenyl-4-triazolecarbonyl chloride, 5-methyl-2-phenyl-2h-1,2,3 triazole-4-carbo nyl chloride, 5-methyl-2-phenyl-2h-1,2,3-triazole-4-carbonylchloride | |
5-methyl-2-phenyltriazole-4-carbonyl chloride |