Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Methyl-2-octanone, tech. 75%, cont. 5-nonanone ca 10%

$16.96 - $363.14

Chemical Identifiers

Chemical Name or Material 5-Methyl-2-octanone
CAS 58654-67-4
Purity Grade Technical Grade
Molecular Formula C9H18O
Formula Weight 142.24
MDL Number MFCD00059414
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2516009

  SDS
alfa aesar™
B2516009
10g Each for $16.96

AAB2516018

  SDS
alfa aesar™
B2516018
50g Each for $43.28

AAB2516030

  SDS
alfa aesar™
B2516030
250g Each for $363.14
Description & Specifications

Specifications

Formula Weight 142.24
TSCA TSCA
CAS 58654-67-4
Molecular Formula C9H18O
Chemical Name or Material 5-Methyl-2-octanone
Purity Grade Technical Grade
Assay Percent Notes cont. 5-nonanone ≈10%
MDL Number MFCD00059414
Quantity 10g
Assay 75%