Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Methyl-2-nitroaniline, 96%

Chemical Identifiers

Chemical Name or Material 5-Methyl-2-nitroaniline
CAS 578-46-1
Molecular Formula C7H8N2O2
Formula Weight 152.15
MDL Number MFCD00007814
UN Number UN2660
Beilstein 2691416
Synonym 3-Amino-4-nitrotoluene; 6-Nitro-m-toluidine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0785206

  SDS
alfa aesar™
L0785206
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL0785214

  SDS
alfa aesar™
L0785214
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 152.15
Chemical Name or Material 5-Methyl-2-nitroaniline
Assay 96%
MDL Number MFCD00007814
CAS 578-46-1
UN Number UN2660
Health Hazard 3 GHS P Statement
P280h-P273-P309-P310-P302+P352
Wear protective gloves/clothing.
Avoid release to the environment.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
Molecular Formula C7H8N2O2
Beilstein 2691416
Synonym 3-Amino-4-nitrotoluene; 6-Nitro-m-toluidine
Melting Point 106° to 110°C
Quantity 5g
Odor Characteristic
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H311-H331-H373
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITROTOLUIDINES (MONO)
RTECS Number XU8060000
Safety and Handling

Danger

GHS P Statement
P280h-P273-P309-P310-P302+P352
Wear protective gloves/clothing.
Avoid release to the environment.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.

GHS H Statement
H301-H311-H331-H373
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.