Please login to your online account to display your discounted pricing

5-Methyl-2-nitroaniline, 95%, Acros Organics


Quantity 1g

Manufacturer: thermo fisher scientific  AC441620010

 View more versions of this product

Catalog No. AC441620010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Chemical Name or Material 5-Methyl-2-nitroaniline
CAS 578-46-1
Assay Percent Range 95%
Molecular Formula C7H8N2O2
Formula Weight 152.15
Quantity 1g

Specifications

Chemical Name or Material 5-Methyl-2-nitroaniline
Assay Percent Range 95%
Molecular Formula C7H8N2O2
Formula Weight 152.15