Please login to your online account to display your discounted pricing

5-Methyl-2-nitroaniline, 95%, Acros Organics

Chemical Identifiers

Chemical Name or Material 5-Methyl-2-nitroaniline
Assay Percent Range 95%
Molecular Formula C7H8N2O2
Formula Weight 152.15
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AC441620010

  SDS
thermo fisher scientific
AC441620010
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC441620050

  SDS
thermo fisher scientific
AC441620050
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material 5-Methyl-2-nitroaniline
CAS 578-46-1
Assay Percent Range 95%
Molecular Formula C7H8N2O2
Formula Weight 152.15
Quantity 1g