Learn More
5-Methyl-2-hexanone, 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 149660010
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
5-Methyl-2-hexanone | |
110-12-3 | |
100.0 | |
Colorless | |
145.0°C | |
Authentic | |
Glass bottle | |
1.4060 to 1.4080 | |
MFCD00008950 | |
0.81 | |
Solubility in water: 5.4g/L (20°C). Other solubilities: miscible with most common organic solvents | |
CC(C)CCC(=O)C | |
114.19 | |
114.19 | |
Extra Pure |
99% | |
98.5 | |
-74.0°C | |
0.8100g/mL | |
41°C | |
98.5% min. (GC) | |
C7H14O | |
(CH3)2CHCH2CH2COCH3 | |
1 L | |
5-methyl-2-hexanone, 2-hexanone, 5-methyl, isoamyl methyl ketone, methyl isoamyl ketone, miak, isopentyl methyl ketone, 2-methyl-5-hexanone, ketone, methyl isoamyl, methyl isopentyl ketone, unii-6o4a4a5f28 | |
FFWSICBKRCICMR-UHFFFAOYSA-N | |
5-methylhexan-2-one | |
8034 | |
99% | |
Liquid |
Chemical Identifiers
110-12-3 | |
114.19 | |
FFWSICBKRCICMR-UHFFFAOYSA-N | |
8034 | |
CC(C)CCC(=O)C |
C7H14O | |
MFCD00008950 | |
5-methyl-2-hexanone, 2-hexanone, 5-methyl, isoamyl methyl ketone, methyl isoamyl ketone, miak, isopentyl methyl ketone, 2-methyl-5-hexanone, ketone, methyl isoamyl, methyl isopentyl ketone, unii-6o4a4a5f28 | |
5-methylhexan-2-one |
Safety and Handling
GHS H Statement:
Harmful if inhaled.
Flammable liquid and vapor.
GHS P Statement:
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
EINECSNumber : 203-737-8
RTECSNumber : MP3850000
TSCA : TSCA
RUO – Research Use Only