Learn More
5-Methyl-2-hexanone, 99%, Thermo Scientific Chemicals
$42.38 - $289.75
Chemical Identifiers
CAS | 110-12-3 |
---|---|
Molecular Formula | C7H14O |
Molecular Weight (g/mol) | 114.188 |
MDL Number | MFCD00008950 |
InChI Key | FFWSICBKRCICMR-UHFFFAOYSA-N |
Synonym | 5-methyl-2-hexanone, 2-hexanone, 5-methyl, isoamyl methyl ketone, methyl isoamyl ketone, miak, isopentyl methyl ketone, 2-methyl-5-hexanone, ketone, methyl isoamyl, methyl isopentyl ketone, unii-6o4a4a5f28 |
PubChem CID | 8034 |
IUPAC Name | 5-methylhexan-2-one |
SMILES | CC(C)CCC(=O)C |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAL17584AE
|
Thermo Scientific Chemicals
L17584AE |
100 mL |
Each for $42.38
|
|
|||||
AAL17584AP
|
Thermo Scientific Chemicals
L17584AP |
500 mL |
Each for $82.23
|
|
|||||
AAL175840F
|
Thermo Scientific Chemicals
L175840F |
2500 mL |
Each for $289.75
|
|
|||||
Description
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.
Solubility
Soluble in water. (5.4 g/L)
Notes
Store in cool dry conditions in well sealed containers. Incompatible with strong oxidizing agents.
Chemical Identifiers
110-12-3 | |
114.188 | |
FFWSICBKRCICMR-UHFFFAOYSA-N | |
8034 | |
CC(C)CCC(=O)C |
C7H14O | |
MFCD00008950 | |
5-methyl-2-hexanone, 2-hexanone, 5-methyl, isoamyl methyl ketone, methyl isoamyl ketone, miak, isopentyl methyl ketone, 2-methyl-5-hexanone, ketone, methyl isoamyl, methyl isopentyl ketone, unii-6o4a4a5f28 | |
5-methylhexan-2-one |
Specifications
110-12-3 | |
0.812 | |
41°C (105°F) | |
1.4075 | |
100 mL | |
506163 | |
Soluble in water. (5.4g/L) | |
CC(C)CCC(=O)C | |
114.188 | |
114.19 | |
5-Methyl-2-hexanone |
-74°C | |
144°C to 145°C | |
C7H14O | |
MFCD00008950 | |
UN2302 | |
5-methyl-2-hexanone, 2-hexanone, 5-methyl, isoamyl methyl ketone, methyl isoamyl ketone, miak, isopentyl methyl ketone, 2-methyl-5-hexanone, ketone, methyl isoamyl, methyl isopentyl ketone, unii-6o4a4a5f28 | |
FFWSICBKRCICMR-UHFFFAOYSA-N | |
5-methylhexan-2-one | |
8034 | |
99% |
Safety and Handling
GHS H Statement
H226-H332
Flammable liquid and vapor.
Harmful if inhaled.
P210-P233-P240-P241-P242-P243-P261-P271-P280-P303+P361+P353-P304+P340-P312-P370+P378q-P501c
H226-H332
DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: 5-METHYLHEXAN-2-ONE
EINECSNumber : 203-737-8
RTECSNumber : MP3850000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only