Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Methyl-2-hexanol, 98%

Chemical Identifiers

Chemical Name or Material 5-Methyl-2-hexanol
CAS 627-59-8
Molecular Formula C7H16O
Formula Weight 116.20g/mol
MDL Number MFCD00004558
UN Number UN1987
Beilstein 1731619
Synonym Methyl isopentyl carbinol; Isopentyl methyl carbinol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2262106

 
alfa aesar™
B22621-06
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB2262114

 
alfa aesar™
B22621-14
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Boiling Point 148° to 150°C
Quantity 5g
Flash Point 46°C (114°F)
Chemical Name or Material 5-Methyl-2-hexanol
CAS 627-59-8
Assay 98%
Molecular Formula C7H16O
Formula Weight 116.20g/mol
MDL Number MFCD00004558
UN Number UN1987
Beilstein 1731619
Synonym Methyl isopentyl carbinol; Isopentyl methyl carbinol
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H226
Flammable liquid and vapour.
Health Hazard 3 GHS P Statement
P210-P280-P240-P303+P361+P353-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ALCOHOLS, N.O.S.
Safety and Handling

Warning

GHS P Statement
P210-P280-P240-P303+P361+P353-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H226
Flammable liquid and vapour.