Learn More
5-Methyl-2-hepten-4-one, predominantly trans, 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals B2233509
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
5-Methyl-2-hepten-4-one | |
81925-81-7 | |
55°C (131°F) | |
1.444 | |
10 g | |
5-methyl-2-hepten-4-one, filbertone, 5-methylhept-2-en-4-one, +/--filbertone, hazelnut ketone, e-5-methyl-2-hepten-4-one, 5-methyl-e-2-hepten-4-one, fema no. 3761, e-5-methylhept-2-en-4-one, 2e-5-methyl-2-hepten-4-one | |
CCC(C)C(=O)C=CC | |
126.199 | |
126.2 |
predominantly trans | |
169°C to 171°C | |
C8H14O | |
MFCD00792509 | |
UN1224 | |
ARJWAURHQDJJAC-GQCTYLIASA-N | |
(E)-5-methylhept-2-en-4-one | |
5362588 | |
99% |
Chemical Identifiers
81925-81-7 | |
126.199 | |
ARJWAURHQDJJAC-GQCTYLIASA-N | |
5362588 | |
CCC(C)C(=O)C=CC |
C8H14O | |
MFCD00792509 | |
5-methyl-2-hepten-4-one, filbertone, 5-methylhept-2-en-4-one, +/--filbertone, hazelnut ketone, e-5-methyl-2-hepten-4-one, 5-methyl-e-2-hepten-4-one, fema no. 3761, e-5-methylhept-2-en-4-one, 2e-5-methyl-2-hepten-4-one | |
(E)-5-methylhept-2-en-4-one |
Safety and Handling
GHS H Statement
H226
Flammable liquid and vapour.
P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
H226-H315-H319-H335
DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: KETONES, LIQUID, N.O.S.
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only