Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Methyl-2-furoyl chloride, 97%

Chemical Identifiers

Chemical Name or Material 5-Methyl-2-furoyl chloride
CAS 14003-11-3
Molecular Formula C6H5ClO2
Formula Weight 144.56
MDL Number MFCD03421416
UN Number UN3261
Synonym 5-Methylfuran-2-carbonyl chloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2769703

  SDS
alfa aesar™
H2769703
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 144.56
Chemical Name or Material 5-Methyl-2-furoyl chloride
Assay 97%
Molecular Formula C6H5ClO2
MDL Number MFCD03421416
CAS 14003-11-3
UN Number UN3261
Synonym 5-Methylfuran-2-carbonyl chloride
Health Hazard 1 Danger
Melting Point 27° to 30°C
Quantity 1g
Flash Point 91°C (196°F)
Health Hazard 2 GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3 GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Recommended Storage 2° to 8°C
DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.

Applications
As pharmaceutical intermediates and reagent. In chemical research. The intermediate 2-halo- acetylfurans (chloro or bromo) were obtained by a hazardous procedure involving the treatment of 2-furoylchloride (highly lachrymatory) with diazomethane followed by the decomposition of the intermediate with dry HCl. Three monoprotic tridentate ligands were synthesized by condensation of 2-furoylchloride with anthranilic acid and β-alanine, and 2-acetylpyridine with orthanilic acid.

Solubility
Reacts with water.

Notes
Highly lachrymatory. Moisture Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, bases, water, moisture.

Safety and Handling

Danger

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

2° to 8°C