missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Methyl-2-(4-methylphenyl)benzimidazole, 95%

$116.19 - $1,384.95

Chemical Identifiers

CAS 7118-66-3
Molecular Formula C15H14N2
Molecular Weight (g/mol) 222.291
MDL Number MFCD00436743
InChI Key OPBSAVHQPYKEQP-UHFFFAOYSA-N
Synonym 1h-benzimidazole, 5-methyl-2-4-methylphenyl, 2-4-methylphenyl-5-methyl-1h-benzimidazole, 5-methyl-2-4-methylphenyl benzimidazole, 5-methyl-2-4-methylphenyl-1h-1,3-benzodiazole, 5-methyl-2-4-methylphenyl-1h-benzimidazole, 5-methyl-2-4-methylphenyl-3h-1,3-benzodiazole, 5-methyl-2-p-tolyl-1h-benzoimidazole, 6-methyl-2-4-methylphenyl-1h-benzimidazole
PubChem CID 893072
IUPAC Name 6-methyl-2-(4-methylphenyl)-1H-benzimidazole
SMILES CC1=CC=C(C=C1)C2=NC3=C(N2)C=C(C=C3)C
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH64908MD View Documents Alfa Aesar™
H64908MD
250mg Each for $116.19
 
AAH6490803 View Documents Alfa Aesar™
H6490803
1g Each for $346.24
 
AAH6490806 View Documents Alfa Aesar™
H6490806
5g Each for $1,384.95
 
Specifications

Chemical Identifiers

7118-66-3
222.291
OPBSAVHQPYKEQP-UHFFFAOYSA-N
893072
CC1=CC=C(C=C1)C2=NC3=C(N2)C=C(C=C3)C
C15H14N2
MFCD00436743
1h-benzimidazole, 5-methyl-2-4-methylphenyl, 2-4-methylphenyl-5-methyl-1h-benzimidazole, 5-methyl-2-4-methylphenyl benzimidazole, 5-methyl-2-4-methylphenyl-1h-1,3-benzodiazole, 5-methyl-2-4-methylphenyl-1h-benzimidazole, 5-methyl-2-4-methylphenyl-3h-1,3-benzodiazole, 5-methyl-2-p-tolyl-1h-benzoimidazole, 6-methyl-2-4-methylphenyl-1h-benzimidazole
6-methyl-2-(4-methylphenyl)-1H-benzimidazole

Specifications

250mg
C15H14N2
1h-benzimidazole, 5-methyl-2-4-methylphenyl, 2-4-methylphenyl-5-methyl-1h-benzimidazole, 5-methyl-2-4-methylphenyl benzimidazole, 5-methyl-2-4-methylphenyl-1h-1,3-benzodiazole, 5-methyl-2-4-methylphenyl-1h-benzimidazole, 5-methyl-2-4-methylphenyl-3h-1,3-benzodiazole, 5-methyl-2-p-tolyl-1h-benzoimidazole, 6-methyl-2-4-methylphenyl-1h-benzimidazole
CC1=CC=C(C=C1)C2=NC3=C(N2)C=C(C=C3)C
222.291
222.29
95%
7118-66-3
MFCD00436743
OPBSAVHQPYKEQP-UHFFFAOYSA-N
6-methyl-2-(4-methylphenyl)-1H-benzimidazole
893072
Solid
5-Methyl-2-(4-methylphenyl)benzimidazole
Safety and Handling

Safety and Handling

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures