Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Methyl-2(3H)-benzoxazolethione, 97%

$44.15 - $44.15

Chemical Identifiers

Chemical Name or Material 5-Methyl-2(3H)-benzoxazolethione
CAS 22876-22-8
Molecular Formula C8H7NOS
Formula Weight 165.21
MDL Number MFCD00052492
Synonym 2-Mercapto-5-methylbenzoxazole; 5-Methylbenzoxazoline-2-thione
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2051503

  SDS
alfa aesar™
B2051503
1g Each for $44.15
Description & Specifications

Specifications

Formula Weight 165.21
Chemical Name or Material 5-Methyl-2(3H)-benzoxazolethione
Assay 97%
Molecular Formula C8H7NOS
MDL Number MFCD00052492
CAS 22876-22-8
Synonym 2-Mercapto-5-methylbenzoxazole; 5-Methylbenzoxazoline-2-thione
Health Hazard 1 Warning
Melting Point 223° to 225°C
Quantity 1g
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.