missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Methyl-2-(2-thienyl)benzimidazole, 95%

$59.51 - $703.20

Chemical Identifiers

CAS 3878-24-8
Molecular Formula C12H10N2S
Molecular Weight (g/mol) 214.286
MDL Number MFCD06312144
InChI Key QIYVOSKFSQXPCG-UHFFFAOYSA-N
Synonym 6-methyl-2-2-thienyl benzimidazole, 5-methyl-2-thiophen-2-yl-3h-1,3-benzodiazole, 5-methyl-2-thiophen-2-yl-1h-benzoimidazole, 5-methyl-2-2-thienyl benzimidazole, 2-5-methylbenzimidazol-2-yl thiophene, 5-methyl-2-thien-2-yl-1h-benzimidazole, 2-2-thienyl-5-methyl-1h-benzimidazole, 5-methyl-2-thiophen-2-yl-1h-benzimidazole, 5-methyl-2-thiophen-2-yl-1h-benzo d imidazole, 6-methyl-2-thiophen-2-yl-1h-benzo d imidazole
PubChem CID 924806
IUPAC Name 6-methyl-2-thiophen-2-yl-1H-benzimidazole
SMILES CC1=CC2=C(C=C1)N=C(N2)C3=CC=CS3
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH64938MD View Documents Alfa Aesar™
H64938MD
250mg Each for $59.51
 
AAH6493803 View Documents Alfa Aesar™
H6493803
1g Each for $175.80
 
AAH6493806 View Documents Alfa Aesar™
H6493806
5g Each for $703.20
 
Specifications

Chemical Identifiers

3878-24-8
214.286
QIYVOSKFSQXPCG-UHFFFAOYSA-N
924806
CC1=CC2=C(C=C1)N=C(N2)C3=CC=CS3
C12H10N2S
MFCD06312144
6-methyl-2-2-thienyl benzimidazole, 5-methyl-2-thiophen-2-yl-3h-1,3-benzodiazole, 5-methyl-2-thiophen-2-yl-1h-benzoimidazole, 5-methyl-2-2-thienyl benzimidazole, 2-5-methylbenzimidazol-2-yl thiophene, 5-methyl-2-thien-2-yl-1h-benzimidazole, 2-2-thienyl-5-methyl-1h-benzimidazole, 5-methyl-2-thiophen-2-yl-1h-benzimidazole, 5-methyl-2-thiophen-2-yl-1h-benzo d imidazole, 6-methyl-2-thiophen-2-yl-1h-benzo d imidazole
6-methyl-2-thiophen-2-yl-1H-benzimidazole

Specifications

250mg
C12H10N2S
6-methyl-2-2-thienyl benzimidazole, 5-methyl-2-thiophen-2-yl-3h-1,3-benzodiazole, 5-methyl-2-thiophen-2-yl-1h-benzoimidazole, 5-methyl-2-2-thienyl benzimidazole, 2-5-methylbenzimidazol-2-yl thiophene, 5-methyl-2-thien-2-yl-1h-benzimidazole, 2-2-thienyl-5-methyl-1h-benzimidazole, 5-methyl-2-thiophen-2-yl-1h-benzimidazole, 5-methyl-2-thiophen-2-yl-1h-benzo d imidazole, 6-methyl-2-thiophen-2-yl-1h-benzo d imidazole
CC1=CC2=C(C=C1)N=C(N2)C3=CC=CS3
214.286
214.29
95%
3878-24-8
MFCD06312144
QIYVOSKFSQXPCG-UHFFFAOYSA-N
6-methyl-2-thiophen-2-yl-1H-benzimidazole
924806
Solid
5-Methyl-2-(2-thienyl)benzimidazole