missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

5-Methyl-2-(2-nitroanilino)-3-thiophenecarbonitrile, TCI America™

$352.92 - $1,226.83

Chemical Identifiers

CAS 138564-59-7
Molecular Formula C12H9N3O2S
Molecular Weight (g/mol) 259.283
MDL Number MFCD06408054
InChI Key NPXUFPFFHANGDL-UHFFFAOYSA-N
Synonym 3-Cyano-5-methyl-2-(2-nitroanilino)thiophene
PubChem CID 395460
IUPAC Name 5-methyl-2-(2-nitroanilino)thiophene-3-carbonitrile
SMILES CC1=CC(=C(S1)NC2=CC=CC=C2[N+](=O)[O-])C#N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M21385G View Documents TCI America
M21385G
5g Each for $352.92
 
M213825G View Documents TCI America
M213825G
25g Each for $1,226.83
 
Specifications

Chemical Identifiers

138564-59-7
259.283
NPXUFPFFHANGDL-UHFFFAOYSA-N
395460
CC1=CC(=C(S1)NC2=CC=CC=C2[N+](=O)[O-])C#N
C12H9N3O2S
MFCD06408054
3-Cyano-5-methyl-2-(2-nitroanilino)thiophene
5-methyl-2-(2-nitroanilino)thiophene-3-carbonitrile

Specifications

Undesignated
5g
C12H9N3O2S
3077
NPXUFPFFHANGDL-UHFFFAOYSA-N
5-methyl-2-(2-nitroanilino)thiophene-3-carbonitrile
395460
Crystal-Powder at 20°C
5-Methyl-2-(2-nitroanilino)-3-thiophenecarbonitrile
111°C
138564-59-7
MFCD06408054
3-Cyano-5-methyl-2-(2-nitroanilino)thiophene
CC1=CC(=C(S1)NC2=CC=CC=C2[N+](=O)[O-])C#N
259.283
259.28
≥97.0% (GC)
Safety and Handling

Safety and Handling

TSCA : No