Please login to your online account to display your discounted pricing

5-Methyl-1H-benzotriazole, 98%, ACROS Organics™


Quantity 50g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  188000500

 View more versions of this product

Catalog No. AC188000500

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Physical Form Crystalline Powder
Formula Weight 133.15
Quantity 50g
Color Cream to beige
TSCA TSCA
RTECS Number DM1400000
EINECS Number 205-265-8
Melting Point 80° to 84°C
Beilstein 26, 58
CAS Max % 100.0
Packaging Glass bottle
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
CAS Min % 97.5
Infrared Spectrum Authentic
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Name Note 98%
Molecular Formula C7H7N3
Assay Percent Range 98%
Chemical Name or Material 5-Methyl-1H-benzotriazole
MDL Number MFCD00005702
CAS 136-85-6

Specifications

Chemical Name or Material 5-Methyl-1H-benzotriazole
Name Note 98%
CAS 136-85-6
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C7H7N3
Formula Weight 133.15
MDL Number MFCD00005702
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause respiratory irritation.

SDS & Safety Documents