Please login to your online account to display your discounted pricing

5-Methyl-1H-benzotriazole, 98%, ACROS Organics™

$43.08 - $164.72

Chemical Identifiers

Chemical Name or Material 5-Methyl-1H-benzotriazole
Name Note 98%
CAS 136-85-6
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C7H7N3
Formula Weight 133.15
MDL Number MFCD00005702
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC188000100

  SDS
thermo fisher scientific
188000100
Glass bottle 10g Each for $43.08

AC188000500

  SDS
thermo fisher scientific
188000500
Glass bottle 50g Each for $164.72
Description & Specifications

Specifications

Physical Form Crystalline Powder
Assay ≥97.5%
Formula Weight 133.15
Quantity 10g
Color Cream to beige
Beilstein 26, 58
EINECS Number 205-265-8
TSCA TSCA
RTECS Number DM1400000
Melting Point 80° to 84°C
CAS Max % 100.0
Chemical Name or Material 5-Methyl-1H-benzotriazole
Packaging Glass bottle
Name Note 98%
Assay Percent Range 98%
Molecular Formula C7H7N3
MDL Number MFCD00005702
CAS 136-85-6
CAS Min % 97.5
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Infrared Spectrum Authentic
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause respiratory irritation.