Learn More
5-Methyl-1H-benzotriazole, 98+%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1061514
Description
5-Methyl-1H-benzotriazole was used in determination of benzothiazoles and benzotriazoles in waste water samples by GC-MS. It is also used as a potential nitrification inhibitor of urea fertilizer in agricultural soils.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications5-Methyl-1H-benzotriazole was used in determination of benzothiazoles and benzotriazoles in waste water samples by GC-MS. It is also used as a potential nitrification inhibitor of urea fertilizer in agricultural soils.
Solubility
Soluble in chloroform, methanol, water (6.0 g/L at 25°C), ethanol, and toluene (slightly).
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications
5-Methyl-1H-benzotriazole | |
77°C to 84°C | |
210°C to 212°C (12 mmHg) | |
C7H7N3 | |
25 g | |
5-methyl-1h-benzotriazole, 5-methylbenzotriazole, tolutriazole, 5-tolyltriazole, 1h-benzotriazole, 5-methyl, 6-methylbenzotriazole, 5-methyl-1h-benzo d 1,2,3 triazole, 5-methyl-1,2,3-benzotriazole, 6-methyl-1,2,3-benzotriazole, t56 bmnnj g1 | |
LRUDIIUSNGCQKF-UHFFFAOYSA-N | |
5-methyl-2H-benzotriazole | |
8705 | |
133.15 |
136-85-6 | |
5.3 | |
Odorless | |
MFCD00005702 | |
116658 | |
Soluble in chloroform,methanol,water (6.0g/L at 25°C),ethanol,and toluene (slightly). | |
CC1=CC2=NNN=C2C=C1 | |
133.15 | |
CHEBI:83455 | |
≥98% |
Chemical Identifiers
136-85-6 | |
133.15 | |
LRUDIIUSNGCQKF-UHFFFAOYSA-N | |
8705 | |
5-methyl-2H-benzotriazole |
C7H7N3 | |
MFCD00005702 | |
5-methyl-1h-benzotriazole, 5-methylbenzotriazole, tolutriazole, 5-tolyltriazole, 1h-benzotriazole, 5-methyl, 6-methylbenzotriazole, 5-methyl-1h-benzo d 1,2,3 triazole, 5-methyl-1,2,3-benzotriazole, 6-methyl-1,2,3-benzotriazole, t56 bmnnj g1 | |
CHEBI:83455 | |
CC1=CC2=NNN=C2C=C1 |
Safety and Handling
GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
EINECSNumber : 205-265-8
RTECSNumber : DM1400000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only