Please login to your online account to display your discounted pricing

5-Methyl-1-phenyl-1H-pyrazole-4-sulfonyl chloride, 95+%, Maybridge


Quantity 5g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  CC15503DE

 View more versions of this product

Catalog No. CC15503DE

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 256.71
Quantity 5g
Assay 95%
Assay Percent Range 95%
Packaging Amber Glass Bottle
Chemical Name or Material 5-Methyl-1-phenyl-1H-pyrazole-4-sulfonyl chloride
Molecular Formula C10H9ClN2O2S
CAS 342405-38-3

Specifications

Chemical Name or Material 5-Methyl-1-phenyl-1H-pyrazole-4-sulfonyl chloride
Assay 95%
Assay Percent Range 95%
Molecular Formula C10H9ClN2O2S
Formula Weight 256.71