missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

5-Methyl-1-phenyl-1H-pyrazole-4-carboxylic acid, 97%, Maybridge

Manufacturer:  Thermo Scientific CC15501DA

 View more versions of this product

Catalog No. CC15501DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

5-Methyl-1-phenyl-1H-pyrazole-4-carboxylic acid
91138-00-0
100.0
C11H10N2O2
USSMIQWDLWJQDQ-UHFFFAOYSA-N
5-methyl-1-phenylpyrazole-4-carboxylic acid
145221
97%
1g
96.5
97%
5-methyl-1-phenyl-1h-pyrazole-4-carboxylic acid, 3-methyl-2-phenylpyrazole-4-carboxylic acid, 3-methyl-2-phenyl-4-pyrazolecarboxylic acid, akos pao-0331, acmc-20am2c, rarechem al be 1317, maybridge1_002413, buttpark 33\08-16, cbi-bb zero/006129, 5-methyl-1-phenyl-1h-pyrazole-4-carboxylicacid
CC1=C(C=NN1C2=CC=CC=C2)C(=O)O
202.213
202.21

Chemical Identifiers

91138-00-0
202.213
5-methyl-1-phenyl-1h-pyrazole-4-carboxylic acid, 3-methyl-2-phenylpyrazole-4-carboxylic acid, 3-methyl-2-phenyl-4-pyrazolecarboxylic acid, akos pao-0331, acmc-20am2c, rarechem al be 1317, maybridge1_002413, buttpark 33\08-16, cbi-bb zero/006129, 5-methyl-1-phenyl-1h-pyrazole-4-carboxylicacid
5-methyl-1-phenylpyrazole-4-carboxylic acid
C11H10N2O2
USSMIQWDLWJQDQ-UHFFFAOYSA-N
145221
CC1=C(C=NN1C2=CC=CC=C2)C(=O)O
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS Documents