missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Methyl-1-phenyl-1H-pyrazole-4-carboxylic acid, 97%

Manufacturer:  Alfa Aesar™ L12594MD

 View more versions of this product

Catalog No. AAL12594MD



Specifications

Specifications

5-Methyl-1-phenyl-1H-pyrazole-4-carboxylic acid
250mg
C11H10N2O2
162776
USSMIQWDLWJQDQ-UHFFFAOYSA-N
5-methyl-1-phenylpyrazole-4-carboxylic acid
145221
97%
165°C to 169°C
91138-00-0
MFCD00067831
5-methyl-1-phenyl-1h-pyrazole-4-carboxylic acid, 3-methyl-2-phenylpyrazole-4-carboxylic acid, 3-methyl-2-phenyl-4-pyrazolecarboxylic acid, akos pao-0331, acmc-20am2c, rarechem al be 1317, maybridge1_002413, buttpark 33\08-16, cbi-bb zero/006129, 5-methyl-1-phenyl-1h-pyrazole-4-carboxylicacid
CC1=C(C=NN1C2=CC=CC=C2)C(=O)O
202.213
202.21

Chemical Identifiers

91138-00-0
202.213
USSMIQWDLWJQDQ-UHFFFAOYSA-N
145221
CC1=C(C=NN1C2=CC=CC=C2)C(=O)O
C11H10N2O2
MFCD00067831
5-methyl-1-phenyl-1h-pyrazole-4-carboxylic acid, 3-methyl-2-phenylpyrazole-4-carboxylic acid, 3-methyl-2-phenyl-4-pyrazolecarboxylic acid, akos pao-0331, acmc-20am2c, rarechem al be 1317, maybridge1_002413, buttpark 33\08-16, cbi-bb zero/006129, 5-methyl-1-phenyl-1h-pyrazole-4-carboxylicacid
5-methyl-1-phenylpyrazole-4-carboxylic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents