Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Methyl-1-phenyl-1H-pyrazole-4-carboxylic acid, 97%

$61.32 - $537.92

Chemical Identifiers

Chemical Name or Material 5-Methyl-1-phenyl-1H-pyrazole-4-carboxylic acid
CAS 91138-00-0
Molecular Formula C11H10N2O2
Formula Weight 202.21
MDL Number MFCD00067831
Beilstein 162776
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1259403

  SDS
alfa aesar™
L1259403
1g Each for $158.22

AAL1259406

  SDS
alfa aesar™
L1259406
5g Each for $537.92

AAL12594MD

  SDS
alfa aesar™
L12594MD
250mg Each for $61.32
Description & Specifications

Specifications

Chemical Name or Material 5-Methyl-1-phenyl-1H-pyrazole-4-carboxylic acid
CAS 91138-00-0
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Assay 97%
Molecular Formula C11H10N2O2
Formula Weight 202.21
MDL Number MFCD00067831
Beilstein 162776
Melting Point 165° to 169°C
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.