missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
5-Methyl-1-phenyl-(1H-pyrazol-4-ylmethyl)amine, 95%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC439760010
Specifications
5-Methyl-1-phenyl-(1H-pyrazol-4-ylmethyl)amine | |
Glass Bottle | |
1g | |
94.0 | |
94% min. (HPLC) | |
1h-pyrazole-4-methanamine, 5-methyl-1-phenyl, 5-methyl-1-phenyl-1h-pyrazol-4-yl methanamine, 5-methyl-1-phenyl-1h-pyrazol-4-yl methyl amine, 5-methyl-1-phenylpyrazol-4-yl methanamine | |
CC1=C(C=NN1C2=CC=CC=C2)CN | |
187.24 | |
187.24 | |
95% |
Authentic | |
Colorless to Yellow or White to Yellow | |
894779-67-0 | |
100.0 | |
C11H13N3 | |
GRLXUIRJMGNTQO-UHFFFAOYSA-N | |
(5-methyl-1-phenylpyrazol-4-yl)methanamine | |
28472963 | |
Liquid or Powder |
Chemical Identifiers
894779-67-0 | |
187.24 | |
1h-pyrazole-4-methanamine, 5-methyl-1-phenyl, 5-methyl-1-phenyl-1h-pyrazol-4-yl methanamine, 5-methyl-1-phenyl-1h-pyrazol-4-yl methyl amine, 5-methyl-1-phenylpyrazol-4-yl methanamine | |
(5-methyl-1-phenylpyrazol-4-yl)methanamine |
C11H13N3 | |
GRLXUIRJMGNTQO-UHFFFAOYSA-N | |
28472963 | |
CC1=C(C=NN1C2=CC=CC=C2)CN |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Causes severe skin burns and eye damage.
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi
GHS Signal Word: Danger