Please login to your online account to display your discounted pricing

5-Methyl-1-phenyl-(1H-pyrazol-4-ylmethyl)amine, 95%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 5-Methyl-1-phenyl-(1H-pyrazol-4-ylmethyl)amine
CAS 894779-67-0
CAS Min % 94.0
CAS Max % 100.0
Assay ≥94.0%
Assay Percent Range 95%
Molecular Formula C11H13N3
Formula Weight 187.24
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AC439760050

 
thermo fisher scientific
AC439760050
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC439760010

 
thermo fisher scientific
AC439760010
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 187.24
Physical Form Liquid or Powder
Assay ≥94.0%
Quantity 5g
Color Colorless to yellow or white to yellow
CAS Min % 94.0
CAS Max % 100.0
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Causes severe skin burns and eye damage.
CAS 894779-67-0
Health Hazard 1 GHS Signal Word: Danger
Chemical Name or Material 5-Methyl-1-phenyl-(1H-pyrazol-4-ylmethyl)amine
Molecular Formula C11H13N3
Assay Percent Range 95%
Infrared Spectrum Authentic
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.
Causes severe skin burns and eye damage.