Learn More
5-Methyl-1-hexyn-3-ol, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H5349014
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
5-Methyl-1-hexyn-3-ol | |
0.86 | |
C7H12O | |
MFCD00041606 | |
UN1987 | |
5-methyl-1-hexyn-3-ol, acmc-209myn, 5-methyl-hex-1-yn-3-ol, 1-hexyn-3-ol,5-methyl, 1-hexyn-3-ol, 5-methyl | |
CC(C)CC(C#C)O | |
112.172 | |
112.17 |
61996-79-0 | |
53°C (127°F) | |
1.436 | |
25 g | |
1741083 | |
NTNUBJHPRAMQPC-UHFFFAOYSA-N | |
5-methylhex-1-yn-3-ol | |
143856 | |
97% |
Chemical Identifiers
61996-79-0 | |
112.172 | |
NTNUBJHPRAMQPC-UHFFFAOYSA-N | |
143856 | |
CC(C)CC(C#C)O |
C7H12O | |
MFCD00041606 | |
5-methyl-1-hexyn-3-ol, acmc-209myn, 5-methyl-hex-1-yn-3-ol, 1-hexyn-3-ol,5-methyl, 1-hexyn-3-ol, 5-methyl | |
5-methylhex-1-yn-3-ol |
Safety and Handling
GHS H Statement
H301-H226
Toxic if swallowed.
Flammable liquid and vapor.
P210-P233-P240-P241-P242-P243-P264b-P270-P280-P301+P312-P303+P361+P353-P330-P370+P378q-P501c
H226-H302
DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ALCOHOLS, N.O.S.
TSCA : No
Recommended Storage : Keep cold
RUO – Research Use Only