missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Methyl-1-hexyn-3-ol, 97%

Manufacturer:  Alfa Aesar™ H5349003

 View more versions of this product

Catalog No. AAH5349003



Specifications

Specifications

5-Methyl-1-hexyn-3-ol
53°C (127°F)
1g
C7H12O
UN1987
1-hexyn-3-ol, 5-methyl
CC(C)CC(C#C)O
112.172
112.17
0.86
1.436
61996-79-0
MFCD00041606
1741083
NTNUBJHPRAMQPC-UHFFFAOYSA-N
5-methylhex-1-yn-3-ol
143856
97%

Chemical Identifiers

61996-79-0
112.172
NTNUBJHPRAMQPC-UHFFFAOYSA-N
143856
CC(C)CC(C#C)O
C7H12O
MFCD00041606
1-hexyn-3-ol, 5-methyl
5-methylhex-1-yn-3-ol
Safety and Handling

Safety and Handling

GHS H Statement
H301-H226
Toxic if swallowed.
Flammable liquid and vapor.

P210-P233-P240-P241-P242-P243-P264b-P270-P280-P301+P312-P303+P361+P353-P330-P370+P378q-P501c

H226-H302

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ALCOHOLS, N.O.S.

TSCA : No

Recommended Storage : Keep cold

Documents