Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Methyl-1-hexene, 99%

Chemical Identifiers

Chemical Name or Material 5-Methyl-1-hexene
CAS 3524-73-0
Molecular Formula (CH3)2CHCH2CH2CH=CH2
Formula Weight 98.19
MDL Number MFCD00039858
UN Number UN3295
Beilstein 1731846
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2071906

 
alfa aesar™
B2071906
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB2071914

 
alfa aesar™
B2071914
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 98.19
Chemical Name or Material 5-Methyl-1-hexene
Assay 99%
Molecular Formula (CH3)2CHCH2CH2CH=CH2
MDL Number MFCD00039858
CAS 3524-73-0
UN Number UN3295
Beilstein 1731846
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H225
Highly flammable liquid and vapour.
Health Hazard 3 GHS P Statement
P210-P280-P240-P303+P361+P353-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: HYDROCARBONS, LIQUID, N.O.S.
Flash Point −10°C (14°F)
Boiling Point 85°C
Quantity 5g
Safety and Handling

Danger

GHS P Statement
P210-P280-P240-P303+P361+P353-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H225
Highly flammable liquid and vapour.